Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
HGF | NM_000601.5 | 13 | 1714 | NP_000592.3 |
NM_001010932.2 | 13 | 1699 | NP_001010932.1 | |
XM_006715956.2 | 14 | 1677 | XP_006716019.1 | |
XM_011516115.2 | 14 | 1662 | XP_011514417.1 | |
AK291674.1 | 13 | 1649 | ||
AK313694.1 | 13 | 1549 | ||
BC130284.1 | 13 | 1562 | ||
BC130286.1 | 13 | 1562 | ||
M29145.1 | 13 | 1588 | AAA52650.1 | |
M55379.1 | 13 | 1472 | ||
M60718.1 | 13 | 1559 | AAA52648.1 | |
M73239.1 | 13 | 1552 | AAA64239.1 | |
M73240.1 | 13 | 1537 | AAA64297.1 | |
X16323.1 | 13 | 1621 | CAA34387.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576398 | Chr7:81685626 - 81732874 on Build GRCh38 | Gain | LOC100128317 HGF |
nsv1019753 | Chr7:81692852 - 82052256 on Build GRCh38 | Gain | LOC101927356 CACNA2D1 HGF |