The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077397.1 | NP_001070865.1 | 1 - 2 | 1032 | 146 |
GenBank mRNA | AY278024.1 | - | 1 - 2 | 1032 | 146 |