The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077503.1 | NP_001070971.1 | 1 - 2 | 102 | 91 |
NM_001077504.1 | NP_001070972.1 | 1 - 2 | 102 | 91 | |
NM_001077505.1 | NP_001070973.1 | 1 - 2 | 102 | 91 | |
NM_001077506.1 | NP_001070974.1 | 1 - 2 | 102 | 91 | |
GenBank mRNA | AF515829.1 | - | 1 - 2 | 102 | 91 |
AF515830.1 | - | 1 - 2 | 102 | 91 | |
AF515831.1 | - | 1 - 2 | 102 | 91 | |
AY471574.1 | - | 1 - 2 | 102 | 91 |