The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077637.1 | NP_001071105.1 | 1 - 1 | 3391 | 85 |
GenBank mRNA | AA843514.1 | - | 1 - 1 | 85 |