The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001255.2 | NP_001246.2 | 10 - 11 | 1427 | 104 |
GenBank mRNA | AF099644.1 | - | 9 - 10 | 1370 | 104 |
AK225504.1 | - | 10 - 11 | 1406 | 104 | |
AK303844.1 | - | 11 - 12 | 1382 | 104 | |
AK312780.1 | - | 10 - 11 | 1406 | 104 | |
BC000624.2 | - | 9 - 10 | 1434 | 104 | |
BC001088.2 | - | 10 - 11 | 1427 | 104 | |
BC006272.2 | - | 9 - 10 | 1369 | 104 | |
BC009425.1 | - | 10 - 11 | 1409 | 104 | |
BC009426.1 | - | 10 - 11 | 1409 | 104 | |
BC010044.2 | - | 10 - 11 | 1418 | 104 | |
BC012803.2 | - | 10 - 11 | 1409 | 104 | |
BC012827.2 | - | 10 - 11 | 1427 | 104 | |
BC013303.2 | - | 10 - 11 | 1409 | 104 | |
BC015998.1 | - | 10 - 11 | 1405 | 104 | |
BC024257.1 | - | 10 - 11 | 1409 | 104 | |
BC031294.1 | - | 10 - 11 | 1409 | 104 | |
BC110321.1 | - | 9 - 10 | 1390 | 104 | |
BT007388.1 | - | 9 - 10 | 1326 | 104 | |
U05340.1 | - | 10 - 11 | 1436 | 104 |