The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002457.3 | NP_002448.3 | - | 3908 | 159 |
GenBank mRNA | L21998.1 | - | - | 3908 | 159 |
M94131.1 | - | 14 - 15 | 2007 | 159 |