Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MUC2
Entrez Gene ID
4583
Gene Name
mucin 2, oligomeric mucus/gel-forming
Gene Aliases
MLP, MUC-2, SMUC
Chromosome Location
Chr.11: 1074875 - 1110508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002457.3 NP_002448.3 - 693 87
GenBank mRNA L21998.1 - - 693 87

View Full Product Details