The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018841.5 | NP_061329.3 | 3 - 4 | 298 | 135 |
XM_017001809.1 | XP_016857298.1 | 3 - 4 | 465 | 135 | |
XM_017001810.1 | XP_016857299.1 | 4 - 5 | 706 | 135 | |
XM_017001811.1 | XP_016857300.1 | 4 - 5 | 635 | 135 | |
GenBank mRNA | AF119663.1 | - | 3 - 4 | 282 | 135 |
AF123766.1 | - | 3 - 4 | 289 | 135 | |
AF365871.1 | - | 3 - 4 | 225 | 135 | |
AF493879.1 | - | 1 - 2 | 93 | 135 | |
AK293101.1 | - | 4 - 5 | 239 | 135 | |
AL832431.1 | - | 3 - 4 | 251 | 135 | |
BC005940.1 | - | 3 - 4 | 135 | ||
BP236225.1 | - | 3 - 4 | 285 | 135 | |
BP236699.1 | - | 3 - 4 | 298 | 135 |