Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYLC1
Entrez Gene ID
1538
Gene Name
cylicin 1
Gene Aliases
CYCL1
Chromosome Location
Chr.X: 83861126 - 83886700 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021118.2 NP_066941.1 2 - 3 121 125
GenBank mRNA BC021669.1 - 2 - 3 105 125
BC058019.1 - 2 - 3 105 125
BC126461.1 - 2 - 3 85 125
DB049397.1 - 2 - 3 121 125

View Full Product Details