Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- RNU2-1
- Entrez Gene ID
-
6066
- Gene Name
- RNA, U2 small nuclear 1
- Gene Aliases
-
RNU2, U2
- Chromosome Location
-
Chr.17: 43233790 - 43233977 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X02665.1 |
- |
- |
|
66 |
View Full Product Details