Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRG2
Entrez Gene ID
448831
Gene Name
FSHD region gene 2
Gene Aliases
FRG2A
Chromosome Location
Chr.4: 190024367 - 190027257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005217.1 NP_001005217.1 4 - 4 1039 113
NM_001286820.1 NP_001273749.1 4 - 4 1042 113
GenBank mRNA AY714545.1 - 4 - 4 1039 113
BC144572.1 - 4 - 4 1027 113

View Full Product Details