The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_015445.1 | 1 - 1 | 149 | ||
GenBank mRNA | AL833948.1 | - | 1 - 1 | 426 | 149 |
AY940073.1 | - | 1 - 1 | 149 | ||
BC132838.1 | - | 1 - 1 | 252 | 149 | |
BC132840.1 | - | 1 - 1 | 252 | 149 |