The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017819.3 | NP_060289.2 | 2 - 2 | 1181 | 159 |
GenBank mRNA | AF226052.1 | - | 2 - 2 | 1149 | 159 |
AK000439.1 | - | 2 - 2 | 1150 | 159 | |
AK026532.1 | - | 1 - 1 | 155 | 159 | |
AY561707.1 | - | 1 - 1 | 983 | 159 | |
BC035967.1 | - | 2 - 2 | 1155 | 159 |