The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000041.3 | NP_000032.1 | 2 - 3 | 155 | 161 |
NM_001302688.1 | NP_001289617.1 | 2 - 3 | 237 | 161 | |
NM_001302689.1 | NP_001289618.1 | 2 - 3 | 86 | 161 | |
NM_001302690.1 | NP_001289619.1 | 2 - 3 | 186 | 161 | |
NM_001302691.1 | NP_001289620.1 | 2 - 3 | 170 | 161 | |
GenBank mRNA | BC003557.1 | - | 2 - 3 | 84 | 161 |
BI550631.1 | - | 2 - 3 | 132 | 161 | |
BM042032.1 | - | 2 - 3 | 170 | 161 | |
BU848796.1 | - | - | 122 | 161 | |
CA771535.1 | - | 1 - 2 | 304 | 161 | |
HY056112.1 | - | 2 - 3 | 155 | 161 | |
HY074384.1 | - | 2 - 3 | 86 | 161 | |
K00396.1 | - | 2 - 3 | 99 | 161 | |
KU177910.1 | - | 1 - 2 | 39 | 161 | |
KU177911.1 | - | 1 - 2 | 39 | 161 | |
KU177912.1 | - | 1 - 2 | 39 | 161 | |
KU177913.1 | - | 1 - 2 | 39 | 161 | |
M12529.1 | - | 2 - 3 | 100 | 161 | |
Z70760.1 | - | 1 - 2 | 76 | 161 |