Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR21
Entrez Gene ID
643905
Gene Name
proline rich 21
Gene Aliases
-
Chromosome Location
Chr.2: 240041813 - 240042982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080835.1 NP_001074304.1 1 - 1 1116 93

View Full Product Details