The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012271.1 | NP_001012271.1 | 3 - 4 | 408 | 157 |
GenBank mRNA | AB028869.1 | - | 3 - 4 | 313 | 157 |
BI520186.1 | - | 3 - 4 | 418 | 157 | |
CB112371.1 | - | 3 - 4 | 341 | 157 | |
DQ508245.1 | - | 3 - 4 | 223 | 161 | |
DQ508246.1 | - | 3 - 4 | 219 | 157 |