The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007104.4 | NP_009035.3 | 3 - 4 | 195 | 92 |
GenBank mRNA | AB082926.1 | - | 1 - 1 | 176 | 92 |
AK313181.1 | - | 3 - 4 | 194 | 92 | |
BC006791.1 | - | 3 - 4 | 176 | 92 | |
BC011366.1 | - | 2 - 3 | 167 | 92 | |
BC070216.1 | - | 1 - 1 | 183 | 92 | |
BC116456.1 | - | 1 - 1 | 128 | 92 | |
BP318268.1 | - | 3 - 4 | 195 | 92 | |
BT020040.1 | - | 2 - 3 | 161 | 92 | |
BT020041.1 | - | 2 - 3 | 161 | 92 | |
U12404.1 | - | 3 - 4 | 176 | 92 |