Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2T4
Entrez Gene ID
127074
Gene Name
olfactory receptor family 2 subfamily T member 4
Gene Aliases
OR1-60, OR2T4Q
Chromosome Location
Chr.1: 248361581 - 248362627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004696.1 NP_001004696.1 1 - 1 949 127

View Full Product Details