The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026767.1 | 2 - 3 | 282 | 65 | |
GenBank mRNA | AK123226.1 | - | 2 - 3 | 282 | 65 |