The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000970.4 | NP_000961.2 | 3 - 4 | 383 | 64 |
NM_001024662.2 | NP_001019833.1 | 3 - 4 | 559 | 64 | |
NM_001320137.1 | NP_001307066.1 | 3 - 4 | 564 | 64 | |
NM_001320138.1 | NP_001307067.1 | 3 - 4 | 598 | 64 | |
NM_001320139.1 | NP_001307068.1 | 3 - 4 | 445 | 64 | |
NM_001320140.1 | NP_001307069.1 | 3 - 4 | 607 | 64 | |
NM_001320141.1 | NP_001307070.1 | 5 - 6 | 672 | 64 | |
NM_001320142.1 | NP_001307071.1 | 3 - 4 | 319 | 64 | |
XM_017019781.1 | XP_016875270.1 | 5 - 6 | 1707 | 64 | |
XM_017019782.1 | XP_016875271.1 | 5 - 6 | 868 | 64 | |
GenBank mRNA | AF261087.1 | - | 1 - 1 | 350 | 64 |
AK311863.1 | - | 3 - 4 | 347 | 64 | |
BC004138.2 | - | 1 - 1 | 359 | 64 | |
BC013863.1 | - | 1 - 1 | 333 | 64 | |
BC020679.1 | - | 1 - 1 | 347 | 64 | |
BC022444.1 | - | 3 - 4 | 514 | 64 | |
BC031009.1 | - | 1 - 1 | 347 | 64 | |
BC032299.1 | - | 1 - 1 | 344 | 64 | |
BC071912.1 | - | 1 - 1 | 336 | 64 | |
BC104824.1 | - | 3 - 4 | 359 | 64 | |
BC104826.1 | - | 3 - 4 | 359 | 64 | |
BG030827.1 | - | 1 - 1 | 109 | 64 | |
BP285025.1 | - | 3 - 4 | 383 | 64 | |
CB140996.1 | - | 3 - 4 | 512 | 64 | |
CD049618.1 | - | 3 - 4 | 529 | 64 | |
CD640506.1 | - | 3 - 4 | 508 | 64 | |
D17554.1 | - | 3 - 4 | 349 | 64 | |
HY016219.1 | - | 3 - 4 | 393 | 64 | |
S71022.1 | - | 2 - 3 | 193 | 64 | |
X69391.1 | - | 3 - 4 | 354 | 64 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK126570.1 | - | - | 1094 | 64 |