The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026542.1 | 1 - 2 | 531 | 145 | |
GenBank mRNA | AF426266.1 | - | 1 - 2 | 150 | 145 |
BC119737.2 | - | 1 - 2 | 124 | 145 | |
BC119738.2 | - | 1 - 2 | 124 | 145 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SH3 domain binding glutamate rich protein | SH3BGR | 21-GARP | -94.1kb | Anti-sense | Chr. - 39445865 - 39515506 on Build GRCh38 |
Downstream | beta-1,3-galactosyltransferase 5 | B3GALT5 | B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5 | 500.0 | Anti-sense | Chr. - 39612940 - 39662889 on Build GRCh38 |