The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000968.3 | NP_000959.2 | 7 - 8 | 903 | 102 |
GenBank mRNA | AB208977.1 | - | 7 - 8 | 880 | 102 |
AK125772.1 | - | 8 - 9 | 1334 | 102 | |
AK130784.1 | - | 7 - 8 | 894 | 102 | |
AK291117.1 | - | 7 - 8 | 912 | 102 | |
AK291859.1 | - | 7 - 8 | 894 | 102 | |
AK297484.1 | - | 2 - 2 | 714 | 102 | |
BC001365.2 | - | 7 - 8 | 884 | 102 | |
BC005817.2 | - | 1 - 1 | 871 | 102 | |
BC007748.2 | - | 1 - 1 | 925 | 102 | |
BC007996.1 | - | 1 - 1 | 877 | 102 | |
BC009888.2 | - | 7 - 8 | 871 | 102 | |
BC010151.2 | - | 7 - 8 | 876 | 102 | |
BC014653.1 | - | 7 - 8 | 872 | 102 | |
BC018840.2 | - | 7 - 8 | 884 | 102 | |
BC040610.1 | - | 1 - 1 | 879 | 102 | |
BC066925.1 | - | 7 - 8 | 894 | 102 | |
BC095427.1 | - | 7 - 8 | 887 | 102 | |
D23660.1 | - | 7 - 8 | 894 | 102 | |
L20868.2 | - | 7 - 8 | 850 | 102 |