The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001864.3 | NP_001855.1 | 2 - 3 | 565 | 85 |
GenBank mRNA | AK312091.1 | - | 2 - 3 | 404 | 85 |
BC002757.2 | - | 2 - 3 | 565 | 85 | |
BT006924.1 | - | 2 - 3 | 103 | 85 | |
CR542120.1 | - | 2 - 3 | 103 | 85 | |
M83186.1 | - | 2 - 3 | 144 | 85 |