Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.X: 119538149 - 119565434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170569.1 NP_001164040.1 6 - 7 769 74
NM_001170570.1 NP_001164041.1 5 - 6 645 74
NM_022101.3 NP_071384.1 6 - 7 687 74
GenBank mRNA AK096297.1 - 6 - 7 733 74
BC023506.2 - 6 - 7 648 74
BQ223940.1 - 6 - 7 652 74

View Full Product Details