Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
Chromosome Location
Chr.15: 68778535 - 68820922 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006305.3 NP_006296.1 3 - 4 494 161
GenBank mRNA AF025684.1 - 3 - 4 471 161
AK127498.1 - 3 - 4 3279 161
AK223280.1 - 3 - 4 440 161
AK312703.1 - 3 - 4 467 161
AY349171.1 - 3 - 4 323 161
BC000608.2 - 3 - 4 458 161
BC007200.1 - 3 - 4 456 161
BC125143.1 - 3 - 4 327 161
BT007436.1 - 3 - 4 323 161
U60823.1 - 3 - 4 323 161
U73477.1 - 3 - 4 419 161
X75090.1 - 3 - 4 426 161

View Full Product Details