Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
Chromosome Location
Chr.15: 68778535 - 68820922 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006305.3 NP_006296.1 5 - 6 790 141
GenBank mRNA AF025684.1 - 5 - 6 767 141
AK127498.1 - 5 - 6 3575 141
AK223280.1 - 5 - 6 736 141
AK312703.1 - 5 - 6 763 141
AY007110.1 - 3 - 4 328 141
AY349171.1 - 5 - 6 619 141
BC007200.1 - 5 - 6 752 141
BT007436.1 - 5 - 6 619 141
U60823.1 - 5 - 6 619 141
U73477.1 - 5 - 6 715 141
X75090.1 - 5 - 6 722 141

View Full Product Details