Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCE3E
Entrez Gene ID
353145
Gene Name
late cornified envelope 3E
Gene Aliases
LEP17
Chromosome Location
Chr.1: 152565654 - 152566770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178435.3 NP_848522.1 2 - 2 328 150
XM_011509484.1 XP_011507786.1 2 - 2 424 150

View Full Product Details