Product Details

Assay Reference Genome
Location

Chr.X:41447497 on build GRCh38
Cytoband
Xp11.4
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
60506
Gene Symbol
NYX
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Location
Chr.X:41447460-41476414 on build GRCh38
Assay Gene Location
Overlaps - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NYX NM_022567.2 NP_072089.1
XM_017029709.1 XP_016885198.1
AF254868.1 AAG42685.1
AJ278865.1 1 64 CAC19014.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv508763 ChrX:41415959 - 41531609 on Build GRCh38 Insertion NYX CASK
esv33720 ChrX:40891487 - 43157518 on Build GRCh38 Loss NYX GPR82 USP9X DDX3X LOC105373185 CASK LOC107985678 LOC643043 PPP1R2P9 GPR34

View Full Product Details