The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002123.4 | NP_002114.3 | 4 - 5 | 855 | 175 |
GenBank mRNA | AB209580.1 | - | 2 - 3 | 3862 | 175 |
AK296613.1 | - | 4 - 5 | 834 | 175 | |
AK316333.1 | - | 4 - 5 | 847 | 175 | |
BQ012862.1 | - | 1 - 2 | 175 |