The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031296.2 | NP_112586.1 | 1 - 2 | 641 | 82 |
XM_011532299.1 | XP_011530601.1 | 2 - 3 | 737 | 82 | |
GenBank mRNA | AF350420.1 | - | 1 - 2 | 363 | 82 |
AK313685.1 | - | 1 - 2 | 301 | 82 | |
AL136904.1 | - | 1 - 2 | 641 | 82 | |
BC036064.1 | - | 1 - 2 | 273 | 82 | |
BC111977.1 | - | 1 - 2 | 293 | 82 |