Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RAB9BP1
- Entrez Gene ID
-
9366
- Gene Name
- RAB9B, member RAS oncogene family pseudogene 1
- Gene Aliases
-
RAB9P1
- Chromosome Location
-
Chr.5: 105099474 - 105100098 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_000039.2 |
|
1 - 1 |
547 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nudix hydrolase 12 |
NUDT12 |
|
-1.5mb
|
Anti-sense |
Chr. - 103548855 - 103562805 on Build GRCh38 |
Downstream |
uncharacterized LOC102467213 |
LOC102467213 |
|
1.7mb
|
Anti-sense |
Chr. - 106815197 - 107011014 on Build GRCh38 |
View Full Product Details