Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SDC4P
- Entrez Gene ID
-
376844
- Gene Name
- syndecan 4 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 30481290 - 30481756 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001580.1 |
|
1 - 1 |
348 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF520746.3 |
- |
1 |
348 |
|
Chr. 22 - 30481290 - 30481756 [-] on Build GRCh38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SEC14 like lipid binding 3 |
SEC14L3 |
TAP2 |
-9.2kb
|
Sense |
Chr. - 30447959 - 30472047 on Build GRCh38 |
Downstream |
SEC14 like lipid binding 4 |
SEC14L4 |
TAP3 |
7.1kb
|
Sense |
Chr. - 30488150 - 30505711 on Build GRCh38 |
View Full Product Details