Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRPC2
- Entrez Gene ID
-
7221
- Gene Name
- transient receptor potential cation channel subfamily C member 2, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 3626460 - 3637559 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002720.2 |
|
6 - 7 |
818 |
93 |
GenBank mRNA |
X89067.1 |
- |
6 - 7 |
801 |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927708 |
LOC101927708 |
|
-64.1kb
|
Anti-sense |
Chr. - 3507980 - 3571987 on Build GRCh38 |
Downstream |
ADP-ribosyltransferase 5 |
ART5 |
ARTC5 |
947.0
|
Anti-sense |
Chr. - 3638503 - 3642392 on Build GRCh38 |
View Full Product Details