Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD88A
- Entrez Gene ID
-
692202
- Gene Name
- small nucleolar RNA, C/D box 88A
- Gene Aliases
-
HBII-180A
- Chromosome Location
-
Chr.19: 50799439 - 50799535 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003067.1 |
|
1 - 1 |
44 |
91 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW444953.1 |
- |
1 |
|
|
Chr. 19 - 50799437 - 50799864 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BF511834.1 |
- |
1 |
|
|
Chr. 19 - 50799437 - 50799888 [+] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 88B |
SNORD88B |
HBII-180B |
-314.0
|
Sense |
Chr. - 50799029 - 50799125 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 88C |
SNORD88C |
HBII-180C |
2.8kb
|
Sense |
Chr. - 50802325 - 50802421 on Build GRCh38 |
View Full Product Details