Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATXN3L
- Entrez Gene ID
-
92552
- Gene Name
- ataxin 3 like
- Gene Aliases
-
MJDL
- Chromosome Location
-
Chr.X: 13318647 - 13320399 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105373133 |
LOC105373133 |
|
-178.0kb
|
Anti-sense |
Chr. - 13044544 - 13086975 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1203 |
LINC01203 |
|
14.8kb
|
Anti-sense |
Chr. - 13335241 - 13341825 on Build GRCh38 |
View Full Product Details