Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAM92A1P2
- Entrez Gene ID
-
403315
- Gene Name
- family with sequence similarity 92, member A3
- Gene Aliases
-
FAM92A3
- Chromosome Location
-
Chr.4: 183037665 - 183040119 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003612.2 |
|
1 - 1 |
2154 |
159 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ429004.1 |
- |
1 |
386 |
|
Chr. 4 - 183039434 - 183040093 [+] on Build GRCh38 |
159 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dCMP deaminase |
DCTD |
|
-120.2kb
|
Anti-sense |
Chr. - 182890091 - 182917477 on Build GRCh38 |
Downstream |
WWC2 antisense RNA 2 |
WWC2-AS2 |
C4orf38 |
56.9kb
|
Anti-sense |
Chr. - 183097021 - 183099199 on Build GRCh38 |
View Full Product Details