The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003675.2 | 1 - 1 | 652 | 144 | |
GenBank mRNA | BC025996.2 | - | 1 - 1 | 652 | 144 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5 | SMARCA5 | ISWI, SNF2H, WCRF135, hISWI, hSNF2H | -2.0kb | Sense | Chr. - 143513463 - 143557489 on Build GRCh38 |
Downstream | FRAS1 related extracellular matrix 3 | FREM3 | 15.9kb | Anti-sense | Chr. - 143577407 - 143700675 on Build GRCh38 |