Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CFHR5 | NM_030787.3 | NP_110414.1 | ||
XM_011510020.2 | XP_011508322.1 | |||
AF295327.1 | AAK15619.1 | |||
AK314026.1 | ||||
BC026282.1 | ||||
BC111773.1 | AAI11774.1 | |||
DA638244.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12e55 | Chr1:196762018 - 197042447 on Build GRCh38 | Gain | CFHR5 F13B CFHR2 CFHR1 CFHR4 CFHR3 |
nsv469599 | Chr1:196827346 - 196986100 on Build GRCh38 | Loss | CFHR5 CFHR2 CFHR1 CFHR4 |
nsv428290 | Chr1:196621707 - 196992672 on Build GRCh38 | Gain+Loss | CFHR5 CFHR2 CFHR1 CFH CFHR4 CFHR3 |
esv3588486 | Chr1:196970045 - 196983172 on Build GRCh38 | Loss | CFHR5 |
nsv471614 | Chr1:196700381 - 196986100 on Build GRCh38 | Gain+Loss | CFHR5 CFHR2 CFHR1 CFH CFHR4 CFHR3 |
esv2721051 | Chr1:196970051 - 196983169 on Build GRCh38 | Deletion | CFHR5 |
nsv528460 | Chr1:196573437 - 197477572 on Build GRCh38 | Gain | CFHR5 KCNT2 F13B CFHR2 CFHR1 ZBTB41 ASPM CFH CRB1 CFHR4 MIR4735 CFHR3 |
nsv8702 | Chr1:196974722 - 196985445 on Build GRCh38 | Loss | CFHR5 |