Product Details

Assay Reference Genome
Location

Chr.1:173912206 on build GRCh38
Cytoband
1q25.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
462
Gene Symbol
SERPINC1
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
Location
Chr.1:173903804-173917378 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SERPINC1 NM_000488.3 NP_000479.1
XM_005245198.3 XP_005245255.1
AF130100.1 AAG35525.1
AK312654.1
BC022309.1 AAH22309.1
D29832.1 BAA06212.1
KP780080.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2758978 Chr1:173841504 - 173986805 on Build GRCh38 Loss SNORD74 RC3H1 SNORD81 GAS5 SNORD75 SNORD44 SNORD79 SNORD77 SNORD80 SNORD47 SNORA103 GAS5-AS1 SNORD78 SNORD76 ZBTB37 DARS2 SERPINC1
nsv831926 Chr1:173733264 - 173925529 on Build GRCh38 Gain+Loss SNORD74 SNORD81 GAS5 KLHL20 SNORD75 SNORD44 SNORD79 SNORD77 SNORD80 SNORD47 CENPL SNORA103 GAS5-AS1 SNORD78 SNORD76 ZBTB37 DARS2 SERPINC1

View Full Product Details