The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000375.2 | NP_000366.1 | 4 - 5 | 514 | 93 |
NM_001324036.1 | NP_001310965.1 | 4 - 5 | 514 | 93 | |
NM_001324037.1 | NP_001310966.1 | 4 - 5 | 514 | 93 | |
NM_001324038.1 | NP_001310967.1 | 4 - 5 | 514 | 93 | |
NM_001324039.1 | NP_001310968.1 | 4 - 5 | 514 | 93 | |
NR_136676.1 | 4 - 5 | 514 | 93 | ||
NR_136677.1 | 4 - 5 | 514 | 93 | ||
XM_005270140.4 | XP_005270197.1 | 4 - 5 | 356 | 93 | |
XM_011540127.1 | XP_011538429.1 | 4 - 5 | 356 | 93 | |
XM_017016610.1 | XP_016872099.1 | 4 - 5 | 356 | 93 | |
XM_017016611.1 | XP_016872100.1 | 4 - 5 | 312 | 93 | |
XM_017016612.1 | XP_016872101.1 | 4 - 5 | 356 | 93 | |
GenBank mRNA | AF230665.1 | - | 3 - 4 | 381 | 93 |
AK314896.1 | - | 4 - 5 | 371 | 93 | |
BC002573.2 | - | 4 - 5 | 473 | 93 | |
J03824.1 | - | 4 - 5 | 444 | 93 | |
KU178377.1 | - | 3 - 4 | 248 | 93 | |
KU178378.1 | - | 3 - 4 | 248 | 93 |