Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UROS
Entrez Gene ID
7390
Gene Name
uroporphyrinogen III synthase
Gene Aliases
UROIIIS
Chromosome Location
Chr.10: 125784980 - 125823268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000375.2 NP_000366.1 8 - 9 833 78
NM_001324036.1 NP_001310965.1 8 - 9 833 78
NM_001324037.1 NP_001310966.1 7 - 8 752 78
NM_001324038.1 NP_001310967.1 7 - 8 752 78
NR_136675.1 6 - 7 652 78
NR_136676.1 8 - 9 833 78
NR_136677.1 8 - 9 833 78
NR_136678.1 5 - 6 563 78
XM_005270140.4 XP_005270197.1 8 - 9 675 78
XM_011540127.1 XP_011538429.1 8 - 9 675 78
XM_017016610.1 XP_016872099.1 8 - 9 675 78
XM_017016611.1 XP_016872100.1 8 - 9 631 78
XM_017016612.1 XP_016872101.1 8 - 9 675 78
GenBank mRNA AF230665.1 - 7 - 8 700 78
AK314896.1 - 8 - 9 690 78
BC002573.2 - 8 - 9 792 78
BI668388.1 - 5 - 6 503 78
J03824.1 - 8 - 9 763 78
KU178377.1 - 7 - 8 567 78
KU178378.1 - 6 - 7 486 78

View Full Product Details