Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4M2
Entrez Gene ID
390538
Gene Name
olfactory receptor family 4 subfamily M member 2
Gene Aliases
OR15-3
Chromosome Location
Chr.15: 22080527 - 22081610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004719.2 NP_001004719.2 1 - 1 552 155
GenBank mRNA BC136985.1 - 1 - 1 552 155

View Full Product Details