The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_002909.2 | 3 - 3 | 2147 | 139 | ||
NR_036473.1 | 4 - 4 | 2255 | 139 | |||
GenBank mRNA | AF075052.1 | - | 1 - 1 | 363 | 139 | |
AJ006835.1 | - | LIT1315 U17HG |
3 - 3 | 2047 | 139 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | small nucleolar RNA, H/ACA box 73B | SNORA73B | 105A, E1c, RNU105A, RNU17B, U17B | -115.0 | Sense | Chr. - 28508558 - 28508715 on Build GRCh38 |
Downstream | tRNA selenocysteine 1 associated protein 1 | TRNAU1AP | PRO1902, SECP43, TRSPAP1 | 42.1kb | Sense | Chr. - 28553017 - 28578545 on Build GRCh38 |