Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location
Chr.10: 121478330 - 121598656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000141.4 NP_000132.3 7 - 7 1540 146
NM_001144915.1 NP_001138387.1 6 - 6 945 146
NM_001144916.1 NP_001138388.1 4 - 4 989 146
NM_001144918.1 NP_001138390.1 5 - 5 1195 146
NM_001320654.1 NP_001307583.1 6 - 6 924 146
NM_001320658.1 NP_001307587.1 7 - 7 1540 146
NM_023029.2 NP_075418.1 5 - 5 776 146
NR_073009.1 5 - 5 1195 294
XM_017015921.1 XP_016871410.1 7 - 7 1738 146
XM_017015923.1 XP_016871412.1 5 - 5 776 146
XM_017015924.1 XP_016871413.1 5 - 5 1393 146
XM_017015925.1 XP_016871414.1 5 - 5 1393 146
GenBank mRNA AK294026.1 - 7 - 7 945 146
AK308621.1 - 4 - 4 989 146
BC037338.2 - 6 - 6 924 146
BC039243.1 - 5 - 5 1195 146
CX873387.1 - 3 - 3 303 146
EF057068.1 - 5 - 5 626 146
EU826598.1 - 6 - 6 893 146
EU826601.1 - 4 - 4 548 146
KP192325.1 - 2 - 2 338 146
KP192326.1 - 5 - 5 710 146
KP192327.1 - 5 - 5 710 146
KP192328.1 - 6 - 6 977 146
KP192329.1 - 4 - 4 632 146
KP192330.1 - 5 - 5 710 146
KP192331.1 - 6 - 6 977 146
M55614.1 - 6 - 6 905 155
M87770.1 - 7 - 7 1166 146
U11814.1 - 7 - 7 1151 294
X52832.1 - 7 - 7 1072 146
Z71929.1 - 6 - 6 905 146

View Full Product Details