Product Details

Assay Reference Genome
Location

Chr.6:21679238 on build GRCh38
Cytoband
6p22.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
401237
Gene Symbol
CASC15
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
Location
Chr.6:21666444-22194400 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CASC15 NR_015410.1
AK022865.1
AK026189.1
AK300802.1
AK310212.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2678811 Chr6:21675625 - 21683263 on Build GRCh38 Deletion CASC15
esv3608312 Chr6:21675564 - 21683456 on Build GRCh38 Loss CASC15

View Full Product Details