Product Details

Assay Reference Genome
Location

Chr.7:100624317 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7036
Gene Symbol
TFR2
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
Location
Chr.7:100620416-100642780 on build GRCh38
Assay Gene Location
Within Intron 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TFR2 NM_001206855.1 NP_001193784.1
NM_003227.3 NP_003218.2
XM_005250553.4 XP_005250610.1
XM_017012573.1 XP_016868062.1
AF067864.1 AAD45561.1
AK000421.1 BAA91153.1
BC142630.1
BC150192.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion PCOLCE-AS1 MOSPD3 MIR4658 AGFG2 NAT16 SLC12A9 ZAN PVRIG MUC17 MOGAT3 GAL3ST4 COPS6 STAG3 SPDYE3 SAP25 TRIP6 PCOLCE GATS PVRIG2P MUC12 AZGP1P1 EPO LAMTOR4 C7orf61 MBLAC1 GNB2 TSC22D4 TAF6 VGF EPHB4 MUC3A FIS1 PILRA TRIM56 MIR93 C7orf43 FBXO24 STAG3L5P ZKSCAN1 ZASP SERPINE1 LRCH4 TFR2 NYAP1 PILRB PLOD3 AP4M1 ZNF3 CNPY4 ZCWPW1 MIR6840 STAG3L5P-PVRIG2P-PILRB MEPCE PPP1R35 UFSP1 POP7 MIR6875 GPC2 PMS2P1 GIGYF1 MIR25 LOC105375429 ACTL6B ZSCAN21 LOC102724094 CLDN15 SRRT ZNHIT1 MIR106B MIR4653 MCM7 AP1S1 ACHE AZGP1
nsv951386 Chr7:100566478 - 100682977 on Build GRCh38 Deletion PCOLCE-AS1 SAP25 MOSPD3 PCOLCE AGFG2 GIGYF1 FBXO24 LOC105375429 GNB2 ZASP ACTL6B LRCH4 TFR2
nsv831072 Chr7:100564880 - 100765212 on Build GRCh38 Loss PCOLCE-AS1 SAP25 MOSPD3 POP7 PCOLCE AGFG2 GIGYF1 FBXO24 EPO LOC105375429 ZAN GNB2 ZASP ACTL6B LRCH4 TFR2

View Full Product Details