Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GOLGA8CP
- Entrez Gene ID
-
729786
- Gene Name
- golgin A8 family member C, pseudogene
- Gene Aliases
-
GOLGA8C
- Chromosome Location
-
Chr.15: 20562349 - 20575703 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027411.2 |
|
11 - 12 |
1530 |
62 |
- Gene Symbol
- GOLGA8DP
- Entrez Gene ID
-
100132979
- Gene Name
- golgin A8 family member D, pseudogene
- Gene Aliases
-
GOLGA8D
- Chromosome Location
-
Chr.15: 23157340 - 23170783 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027407.1 |
|
11 - 12 |
1530 |
62 |
GenBank mRNA |
BC110417.1 |
- |
11 - 12 |
1501 |
62 |
- Gene Symbol
- GOLGA8EP
- Entrez Gene ID
-
390535
- Gene Name
- golgin A8 family member E, pseudogene
- Gene Aliases
-
GOLGA8E
- Chromosome Location
-
Chr.15: 22424673 - 22438026 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033350.1 |
|
11 - 12 |
1530 |
62 |
GenBank mRNA |
BC063309.1 |
- |
11 - 12 |
1504 |
62 |
DQ309037.1 |
- |
11 - 12 |
1504 |
62 |
- Gene Symbol
- GOLGA8F
- Entrez Gene ID
-
100132565
- Gene Name
- golgin A8 family member F
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 28378638 - 28391266 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033351.1 |
|
11 - 12 |
1530 |
62 |
GenBank mRNA |
AK302625.1 |
- |
12 - 13 |
1231 |
62 |
BC144672.1 |
- |
9 - 10 |
1227 |
62 |
BC150302.1 |
- |
11 - 12 |
1504 |
62 |
- Gene Symbol
- GOLGA8G
- Entrez Gene ID
-
283768
- Gene Name
- golgin A8 family member G
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 28519611 - 28532997 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033353.1 |
|
11 - 12 |
1530 |
62 |
GenBank mRNA |
BC099913.1 |
- |
10 - 11 |
2357 |
62 |
BC125193.1 |
- |
10 - 11 |
1322 |
62 |
BC125194.1 |
- |
10 - 11 |
1322 |
62 |
- Gene Symbol
- GOLGA8S
- Entrez Gene ID
-
653061
- Gene Name
- golgin A8 family member S
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 23354748 - 23368324 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038843.1 |
|
11 - 12 |
1483 |
62 |
View Full Product Details