Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
11082
Gene Name
endothelial cell specific molecule 1
Gene Aliases
endocan
Chromosome Location
Chr.5: 54977867 - 54985586 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135604.1 NP_001129076.1 1 - 2 366 102
GenBank mRNA AJ973643.1 - 1 - 2 313 102
AK300634.1 - 1 - 2 373 102

View Full Product Details