Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW383156.1 |
- |
1 - 2 |
314 |
|
Chr. 8 - 12387763 - 12393525 [-] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
family with sequence similarity 66 member A |
FAM66A |
|
|
Anti-sense |
Chr. - 12362019 - 12411001 on Build GRCh38 |
Downstream |
family with sequence similarity 90, member A7 pseudogene |
FAM90A25P |
|
21.0kb
|
Sense |
Chr. - 12414521 - 12418032 on Build GRCh38 |
View Full Product Details