Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100130456
Entrez Gene ID
100130456
Gene Name
uncharacterized LOC100130456
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK095012.1 - 1 - 1 1850 90


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream G protein-coupled receptor 146 GPR146 PGR8 Sense Chr. - 1044573 - 1059269 on Build GRCh38
Downstream G protein-coupled estrogen receptor 1 GPER1 CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER 34.8kb Sense Chr. - 1086807 - 1093815 on Build GRCh38

View Full Product Details